missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine, 98%, Thermo Scientific™

Product Code. p-7047724
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11375495 250 mg 250mg
11365495 1 g 1g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11375495

Brand: Thermo Scientific Alfa Aesar L19680.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31886-57-4
Molecular Formula C14H19FeN
Molecular Weight (g/mol) 257.16
MDL Number MFCD00066272
IUPAC Name (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine
SMILES [Fe].c1cccc1.C[C@H](N(C)C)c1cccc1

Specifications

Density 1.222
Boiling Point 120°C to 121°C (0.7 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.589
Quantity 250 mg
Optical Rotation −13° (c=1 in Ethanol, 5H)
Formula Weight 257.16
Percent Purity 98%
Chemical Name or Material (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.