Learn More
S-Methyl thiobutyrate, 98%
Product Code. p-7026239
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11429667 5 g 5g
11439667 25 g 25g
2 options
Product Code. 11429667
Brand: Thermo Scientific Alfa Aesar B22144.06
Request Bulk or Custom Format
CAS: 2432-51-1 | C5H10OS | 118.194 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2432-51-1
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00009872
InChI Key GRLJIIJNZJVMGP-UHFFFAOYSA-N
Synonym s-methyl thiobutyrate, butanethioic acid, s-methyl ester, methyl thiolbutyrate, methanethiol butyrate, methanethiol n-butyrate, butyric acid, thio-, s-methyl ester, s-methyl thiobutanoate, unii-2p1e432myz, methyl thiobutanoate, methyl thiobutyrate natural
PubChem CID 62444
IUPAC Name S-methyl butanethioate
SMILES CCCC(=O)SC
Specifications
Density 0.966
Boiling Point 142°C to 143°C
Flash Point 34°C (93°F)
Refractive Index 1.462
Quantity 5 g
UN Number UN1993
Beilstein 1848987
Formula Weight 118.2
Percent Purity 98%
Chemical Name or Material S-Methyl thiobutyrate
compliance-icons compliance-icons
Product Identifier
  • S-Methyl thiobutyrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : S-Methyl thiobutyrate