Learn More
(S)-(+)-Lysine Monohydrochloride, Sigma-Aldrich™
Product Code. 15820221
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15820221 500 g 500g
15810221 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15820221 Supplier Sigma-Aldrich Supplier No. 8160180500
 Restricted Item
Estimated Shipment: 12-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 657-27-2
Grade Research
Molecular Weight (g/mol) 182.65
Specifications
Melting Point 262°C
MDL Number MFCD00064564
Synonym (S)-(+)-Lysine monohydrochloride, Lys, (S)-(+)-Lysine monohydrochloride
Density 1.28 g/cm3 at 20°C
Quantity 500 g
Chemical Name or Material (S)-(+)-Lysine Monohydrochloride