Learn More
Invitrogen™ S. Cerevisiae Hansen BY4743
Product Code. 10612384
Change view
Click to view available options
Product Type:
S. Cerevisiae Hansen BY4743
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10612384 S. Cerevisiae Hansen BY4743 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10612384 Supplier Invitrogen™ Supplier No. 95400.BY4743
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The S. cerevisiae Hansen strains are the parental yeast strains from which the deletions were derived.
The S. cerevisiae Hansen strains are the parental yeast strains from which the deletions were derived.
TRUSTED_SUSTAINABILITY
Specifications
Product Type S. Cerevisiae Hansen BY4743
Content And Storage Store in ultra-cold freezer (-68°C to -85°C).