Learn More
Heidolph Instruments S-Box Package 2
Product Code. 17838000 Shop All Heidolph Instruments Products
missing translation for 'orderingAttributeHoverText'
Color:
Blue
Green
Orange
Violet
missing translation for 'unitSize'
Each
Product Code. 17838000
missing translation for 'mfr': Heidolph Instruments 4000300300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The S-Box Package 2 includes additional accessories to complete an existing S-Biosystem incubation system.
  • Each S-box is a stand-alone miniature incubator.
  • Inside the S-box there is room for one standard multi-well plate, or one cell culture bottle (up to T75).
  • The complete interior (inner box) can be purchased as a sterilized exchangeable element in a sterile outer packaging.
  • Your culture is prevented from drying out by a sachet of sterilized water (Waterbag), which is enough for weeks.
  • To avoid mix-up, the front cover of the S-box is available in different colors.
TRUSTED_SUSTAINABILITY
Specifications
Color Violet
Includes 1 S-box, 1 tool kit, 8 inner boxes, 8 water bags, 4 front covers