missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Bay-K-8644, MedChemExpress

Product Code. p-200101529
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. Quantity unitSize
16476318 10 mg 10mg
16432437 50 mg 50mg
16428907 5 mg 5mg
3 options
This item is not returnable. View return policy

Product Code. 16476318

Brand: Medchem Express HY15124

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-(-)-Bay-K-8644 is an agonist of L-type Ca2+ channel. (S)-(-)-Bay-K-8644 activates Ba2+ currents (IBa) (EC50=32 nM).

(S)-(-)-Bay-K-8644 is an agonist of L-type Ca2+ channel. (S)-(-)-Bay-K-8644 activates Ba2+ currents (IBa) (EC50=32 nM).

Specifications

Chemical Name or Material (S)-(-)-Bay-K-8644
CAS 98625-26-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H15F3N2O4
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (350.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC1=C([N+]([O-])=O)[C@@H](C2=C(C(F)(F)F)C=CC=C2)C(C(OC)=O)=C(C)N1
Molecular Weight (g/mol) 356.3
Formula Weight 356.3
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.