Learn More
(S)-(+)-Alanine, Sigma-Aldrich™
Product Code. 15840191
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15840191 100 g 100g
15830191 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15840191 Supplier Sigma-Aldrich Supplier No. 8160000100
 Restricted Item
Estimated Shipment: 12-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56-41-7
Grade Research
Molecular Weight (g/mol) 89.09
Specifications
Melting Point 295°C to 297°C (Decomposition)
MDL Number MFCD00166369
Synonym (S)-(+)-Alanine, 2-Aminopropionic acid, Ala, (S)-(+)-Alanine
Density 1.40 g/cm3 at 20°C
Quantity 100 g
Chemical Name or Material (S)-(+)-Alanine