Learn More
S-Adenosyl-L-methionine disulfate tosylate, MedChemExpress
Description
S-Adenosyl-L-methionine disulfate tosylate (Ademetionine disulfate tosylate) is the principal biological methyl donor synthesized in all mammalian cells but most abundantly in the liver.
Specifications
Specifications
| Chemical Name or Material | S-Adenosyl-L-methionine disulfate tosylate |
| CAS | 97540-22-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H34N6O16S4 |
| Synonym | Ademetionine disulfate tosylateS-Adenosyl methionine disulfate tosylateAdoMet disulfate tosylate |
| Solubility Information | DMSO : ≥ 50 mg/mL (65.21 mM) ∣H2O : 33.33 mg/mL (43.47 mM; Need ultrasonic) |
| SMILES | N[C@@H](CC[S+](C[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3N)O1)O)O)C)C(O)=O.O=S(O)([O-])=O.O=S(O)(O)=O.OS(=O)(C4=CC=C(C)C=C4)=O |
| Molecular Weight (g/mol) | 766.8 |
| Quantity | 500 mg |
| Show More |