Learn More
(S)-Adamantylglycine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (S)-Adamantylglycine Hydrochloride |
| CAS | 102502-64-7 |
| Molecular Formula | C12H20ClNO2 |
| Quantity | 1 g |
| Synonym | (αS)-Amino-tricyclo[3.3.1.13,7]decane-1-acetic acid Hydrochloride |
| InChI Formula | InChI=1S/C12H18NO2.ClH/c13-10(11(14)15)12-4-7-1-8(5-12)3-9(2-7)6-12;/h7-8,10H,1-6,13H2,(H,14,15);1H/t7-,8+,10-,12-;/m1./s1 |
| SMILES | N[C@H](C(O)=O)C1(C[C@H]2C3)C[C@H]3C[C@H](C2)C1.Cl |
| IUPAC Name | (S)-2-((3S,5S,7S)-adamantan-1-yl)-2-aminoacetic acid hydrogen chloride salt |
| Molecular Weight (g/mol) | 245.75 |
| Formula Weight | 245.12 |