Learn More
(S)-4-Fluorophenylglycine, TRC
Specifications
Specifications
| Chemical Name or Material | L-4-Fluorophenylglycine |
| CAS | 19883-57-9 |
| Molecular Formula | C8 H8 F N O2 |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C8H8FNO2/c9-6-3-1-5(2-4-6)7(10)8(11)12/h1-4,7H,10H2,(H,11,12)/t7-/m0/s1 |
| SMILES | N[C@H](C(=O)O)c1ccc(F)cc1 |
| IUPAC Name | (2S)-2-amino-2-(4-fluorophenyl)acetic acid |
| Molecular Weight (g/mol) | 169.15 |
| Formula Weight | 169.0539 |