Learn More
S 3304, TRC
Specifications
Specifications
| Chemical Name or Material | N-[(5-[2-(4-Methylphenyl)ethynyl]-2-thienyl)sulfonyl]-D-tryptophan |
| CAS | 203640-27-1 |
| Molecular Formula | C24H20N2O4S2 |
| Quantity | 10 mg |
| Synonym | S 3304, N-[[5-[2-(4-Methylphenyl)ethynyl]-2-thienyl]sulfonyl]-D-tryptophan |
| InChI Formula | InChI=1S/C24H20N2O4S2/c1-16-6-8-17(9-7-16)10-11-19-12-13-23(31-19)32(29,30)26-22(24(27)28)14-18-15-25-21-5-3-2-4-20(18)21/h2-9,12-13,15,22,25-26H,14H2,1H3,(H,27,28)/t22-/m1/s1 |
| SMILES | Cc1ccc(cc1)C#Cc2ccc(s2)S(=O)(=O)N[C@H](Cc3c[nH]c4ccccc34)C(=O)O |
| IUPAC Name | (2R)-3-(1H-indol-3-yl)-2-[[5-[2-(4-methylphenyl)ethynyl]thiophen-2-yl]sulfonylamino]propanoic acid |
| Molecular Weight (g/mol) | 464.56 |
| Formula Weight | 464.0864 |