Learn More
(S)-3-(Boc-amino)-4-phenylbutyric acid, 95%
Product Code. 15408727
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15408727 250 mg 250mg
15498717 1 g 1g
2 options
Product Code. 15408727
Brand: Thermo Scientific Alfa Aesar H52174.MD
Request Bulk or Custom Format
CAS: 51871-62-6 | C15H21NO4 | 279.34 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51871-62-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD01076271
InChI Key ACKWQHCPHJQANL-LBPRGKRZSA-N
Synonym s-3-boc-amino-4-phenylbutyric acid, boc-l-beta-homophenylalanine, boc-beta-hophe-oh, 3s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, s-3-tert-butoxycarbonyl amino-4-phenylbutanoic acid, 3r-3-tert-butoxy carbonyl amino-4-phenylbutanoic acid, boc-hophe-oh, pubchem12107, tmba023, 3s-n-boc-3-benzyl-beta-alanine
PubChem CID 2761538
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC(O)=O)CC1=CC=CC=C1
Specifications
Melting Point 101°C to 107°C
Beilstein 3060457
Quantity 250 mg
Formula Weight 279.34
Optical Rotation −19.0° (c=1 in Dichloromethane)
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-4-phenylbutyric acid