Learn More
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid, 98+%
Product Code. p-7045715
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372577 25 g 25g
11382577 100 g 100g
11392577 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11372577 Supplier Thermo Scientific Alfa Aesar Supplier No. L11711.14
Add to basket
CAS: 98-79-3 | C5H7NO3 | 129.12 g/mol
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid is used as a building block for pharmaceuticals and asymmetric synthesis. It Is also used as a resolving agent for chiral bases and in an efficient synthesis of quinolizidine and indolizidine alkaloids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid is used as a building block for pharmaceuticals and asymmetric synthesis. It Is also used as a resolving agent for chiral bases and in an efficient synthesis of quinolizidine and indolizidine alkaloids.
Solubility
Soluble in water (10-15% at 20°C), alcohol, acetone, dimethyl sufoxide and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98-79-3
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00005272
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxo
PubChem CID 7405
ChEBI CHEBI:18183
SMILES OC(=O)C1CCC(=O)N1
Specifications
Melting Point 158°C to 162°C
Quantity 25 g
Beilstein 82132
Merck Index 14,8001
Solubility Information Soluble in water (10-15% at 20°C),alcohol,acetone,dimethyl sufoxide and methanol.
Optical Rotation −10.5° (c=5 in Water)
IUPAC Name (2S)-5-oxopyrrolidine-2-carboxylic acid
Formula Weight 129.12
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-2-Pyrrolidinone-5-carboxylic acid