missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylglycinol, 98+%

Product Code. 11368006
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11368006 5 g 5g
11358006 1 g 1g
11378006 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11368006

Brand: Thermo Scientific Alfa Aesar L13265.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20989-17-7 | C8H11NO | 137.182 g/mol

(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.

Solubility
Soluble in chloroform, dimethylsulfoxide and methanol. Limited solubility in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with oxidizing agents. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20989-17-7
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00064404
InChI Key IJXJGQCXFSSHNL-MRVPVSSYSA-N
Synonym s-+-2-phenylglycinol, s-2-phenylglycinol, s-2-amino-2-phenylethanol, s-phenylglycinol, h-phenylglycinol, 2s-2-amino-2-phenylethanol, 2s-2-amino-2-phenylethan-1-ol, l-2-phenylglycinol, h-phg-ol
PubChem CID 134797
IUPAC Name (2S)-2-amino-2-phenylethanol
SMILES C1=CC=C(C=C1)C(CO)N

Specifications

Melting Point 76°C to 78°C
Quantity 5 g
Beilstein 3196190
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,dimethylsulfoxide and methanol. Limited solubility in water.
Optical Rotation +30° (c=0.75 in 1N HCl)
Formula Weight 137.18
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-2-Phenylglycinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.