Learn More
(S)-(+)-2-Octanol, 99%
Product Code. 11308805
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308805 5 g 5g
11398795 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11308805 Supplier Thermo Scientific Alfa Aesar Supplier No. L12425.06
Request Bulk or Custom Format
CAS: 6169-06-8 | C8H18O | 130.23 g/mol
(S)-(+)-2-Octanol is used as chiral compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-(+)-2-Octanol is used as chiral compounds.
Solubility
Soluble in water. (1.12 g/L) at 25°C
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6169-06-8
MDL Number MFCD00064283
Specifications
Density 0.819
Boiling Point 179°C to 180°C
Flash Point 76°C (169°F)
Odor Pleasant
Molecular Formula C6H9O6Pr
Refractive Index 1.426
Quantity 5 g
Beilstein 1719323
Merck Index 14,6752
Solubility Information Soluble in water. (1.12g/L) at 25°C
InChI Key KPXRPRLCONABEG-UHFFFAOYSA-K
Optical Rotation +10° (c = 1 in Ethanol)
SMILES [Pr+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
IUPAC Name praseodymium(3+) triacetate
Molecular Weight (g/mol) 318.04
Formula Weight 130.23
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-Octanol