Learn More
(S)-(+)-2-Amino-1-propanol, 98%
Product Code. 11411407
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11411407 5 g 5g
11401407 1 g 1g
11421407 25 g 25g
3 options
Product Code. 11411407
Brand: Thermo Scientific Alfa Aesar B24916.06
Request Bulk or Custom Format
CAS: 2749-11-3 | C3H10NO | 76.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2749-11-3
MDL Number MFCD00064412
ChEBI CHEBI:78502
Specifications
Density 0.965
Boiling Point 174°C to 176°C
Flash Point 83°C (181°F)
Molecular Formula C3H10NO
Refractive Index 1.4498
Quantity 5 g
UN Number UN2735
Beilstein 1718865
Sensitivity Air sensitive; Hygroscopic
Synonym l-alaninol, s-+-2-amino-1-propanol, s-2-aminopropan-1-ol, 2s-2-aminopropan-1-ol, h-alaninol, alaninol, s-2-amino-1-propanol, +-2-aminopropan-1-ol, 2-aminopropanol, +, unii-v403gh89l1
InChI Key BKMMTJMQCTUHRP-VKHMYHEASA-O
Optical Rotation +22° (c=2 in Ethanol)
SMILES CC(CO)N
IUPAC Name (2S)-2-aminopropan-1-ol
Molecular Weight (g/mol) 76.12
PubChem CID 80307
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (S)-(+)-2-Amino-1-propanol