Learn More
(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine, 97+%, Thermo Scientific Chemicals
Product Code. p-7040662
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15445796
Brand: Thermo Scientific Alfa Aesar H60488.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine is used as pharmaceutical intermediates.
Notes
Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 607389-84-4
Molecular Formula C24H24FeNP
Molecular Weight (g/mol) 413.28
MDL Number MFCD11045439
InChI Key DRBVPPSGNFMTKZ-JEZYAYIFNA-N
Synonym (Rp)-1-[(1S)-(1-Aminoethyl)]-2-(diphenylphosphino)ferrocene
PubChem CID 131675344
IUPAC Name cyclopenta-1,3-diene;(1S)-1-(2-diphenylphosphanylcyclopenta-2,4-dien-1-yl)ethanamine;iron
SMILES [Fe].c1cccc1.C[C@H](N)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 131°C to 135°C
Quantity 250 mg
Formula Weight 413.27
Percent Purity ≥97%
Chemical Name or Material (S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine