missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Ferrocenylethanol, 99%, Thermo Scientific™

Product Code. p-7047727
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11315505

Brand: Thermo Scientific Alfa Aesar L19683.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33136-66-2
Molecular Formula C12H14FeO
Molecular Weight (g/mol) 230.09
MDL Number MFCD01074672
IUPAC Name (S)-1-Ferrocenylethanol
SMILES [Fe].c1cccc1.C[C@H](O)c1cccc1

Specifications

Melting Point 77°C to 79°C
Quantity 250 mg
Formula Weight 230.09
Percent Purity 99%
Chemical Name or Material (S)-1-Ferrocenylethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.