Learn More
(S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 86069-86-5 |
| Molecular Formula | C21H21NO4 |
| MDL Number | MFCD00235898 |
| Quantity | 5 g |
| SMILES | O=C(O)[C@H]1N(C(OCC2C3=CC=CC=C3C4=CC=CC=C24)=O)CCCC1 |
| Molecular Weight (g/mol) | 351.40 |
| Percent Purity | 98% |
| Chemical Name or Material | (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid |