Learn More
(S)-1,8-Dihydroxy-3-(hydroxymethyl)-10-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)anthracen-9(10H)-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1415-73-2 |
| Molecular Formula | C21H22O9 |
| MDL Number | MFCD00151160 |
| Quantity | 5 g |
| SMILES | O=C1C2=C(C=CC=C2O)[C@H]([C@H]3[C@@H]([C@H]([C@@H]([C@@H](CO)O3)O)O)O)C4=CC(CO)=CC(O)=C14 |
| Molecular Weight (g/mol) | 418.39 |
| Percent Purity | 98% |
| Chemical Name or Material | (S)-1,8-Dihydroxy-3-(hydroxymethyl)-10-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)anthracen-9(10H)-one |