Learn More
(S)-1-(4-Nitrophenyl)ethanamine Hydrochloride, TRC
Product Code. 30466247
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466247 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30466247 Supplier Toronto Research Chemicals Supplier No. TRCN5452802.5G
 Restricted Item
Estimated Shipment: 08-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (S)-1-(4-Nitrophenyl)ethanamine Hydrochloride
CAS 132873-57-5
Molecular Formula C8H10N2O2.HCl
Quantity 2.5 g
Synonym (1S)-1-(4-Nitrophenyl)ethan-1-amine Hydrochloride, (S)-(-)-α-Methyl-4-nitrobenzylamine Hydrochloride, (S)-α-Methyl-4-nitrobenzylamine Hydrochloride, 1-(S)-(4-Nitrophenyl)ethylamine Hydrochloride, [(S)-1-(4-Nitrophenyl)ethyl]amine Hydrochloride
InChI Formula InChI=1S/C8H10N2O2.ClH/c1-6(9)7-2-4-8(5-3-7)10(11)12;/h2-6H,9H2,1H3;1H/t6-;/m0./s1
SMILES C[C@H](N)C1=CC=C([N+]([O-])=O)C=C1.Cl
IUPAC Name (S)-1-(4-nitrophenyl)ethan-1-amine hydrochloride
Molecular Weight (g/mol) 202.63
Formula Weight 202.0509