Learn More
IKA™ RV 10.601 Model Distilling Spider
Product Code. 11825922 Shop All IKA Products
Change view
Click to view available options
Quantity:
1 Pack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11825922 1 Pack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11825922 Supplier IKA™ Supplier No. 0003739500
  • ONLINE EXCLUSIVE PRICE
    15585.00 NOK
    14014.53 NOK / Pack of 1
    Save 1570.47 NOK
    10% Off
In Stock
Specifications
Type Distilling Spider
For Use With (Application) For simultaneous distillation in 12 Distilling sleeves
Quantity 1 Pack
Standard Taper Joint Side NS 29/32
Volume (Metric) 20 mL