Learn More
IKA™ RV 10.600 Model Distilling Spider
Product Code. 11815922 Shop All IKA Products
Click to view available options
Quantity:
1 Pack
Unit Size:
Each
Product Code. 11815922
Brand: IKA™ 0003739400
Only null left
Specifications
Type Distilling Spider
For Use With (Application) For simultaneous distillation in 6 Distilling sleeves
Quantity 1 Pack
Standard Taper Joint Side NS 29/32
Volume (Metric) 20 mL