Learn More
RUNX3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27099-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16885385
  • 1611.84 NOK / 20µL
Estimated Shipment: 24-05-2024
Description
The RUNX3 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human RUNX3. This antibody recognizes human, mouse antigen. The RUNX3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RUNX3
Polyclonal
Unconjugated
20 μL
864
IgG
Western Blot, ELISA
367 μg/mL
Rabbit
Primary
Human, Mouse