Learn More
RUNX3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27099-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The RUNX3 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human RUNX3. This antibody recognizes human, mouse antigen. The RUNX3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
RUNX3 | |
Polyclonal | |
Unconjugated | |
150 μL | |
864 | |
IgG |
Western Blot, ELISA | |
367 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |