missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RUNX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568508
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
RUNX2, a nuclear protein, is a member of the RUNX family of transcription factors and has an Runt DNA-binding domain. It is essential for membranous and endochondral bone formation. It not only regulates osteoblastic differentiation and skeletal morphogenesis but also acts as a scaffold for nucleic acids and regulatory factors involved in skeletal gene expression. The protein can bind DNA both as a monomer and with more affinity, as a subunit of a heterodimeric complex. It plays a critical role in increasing TGFBR1 expression by osteoblasts and functions in cooperation with DLX5 or a related factor to activate osteoblast-specific gene expression. Mutations lead to disorders in bone development like cleidocranial dysplasia (CCD).
Specifications
| RUNX2 | |
| Polyclonal | |
| Unconjugated | |
| RUNX2 | |
| Acute myeloid leukemia 3 protein; AKV core binding factor; Aml3; AML-3; Cbf; Cbfa1; Cbfa-1; CBF-alpha-1; CCD; CCD1; CLCD; core binding factor alpha 1; core binding factor alpha 1 subunit; core-binding factor subunit alpha-1; core-binding factor, runt domain, alpha subunit 1; LOW QUALITY PROTEIN: runt-related transcription factor 2; LS3; Oncogene AML-3; Osf2; OSF-2; osteoblast-specific transcription factor 2; PEA2aA; PEA2-alpha A; PEBP2 alpha A; PEBP2A; Pebp2a1; PEBP2aA; PEBP2-alpha A; Pebpa2a; Polyomavirus enhancer-binding protein 2 alpha A subunit; runt domain, alpha subunit 1; runt related transcription factor 2; runt-related transcription factor 2; RUNX 2; RUNX family transcription factor 2; Runx2; RUNX-2; SL3/AKV core-binding factor alpha A subunit; SL3-3 enhancer factor 1 alpha A subunit; transcription factor RUNX2; transcription factor RUNX2/CBFA1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 367218 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Z2J9 | |
| RUNX2 | |
| synthetic peptide corresponding to a region of rat RUNX2. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction