Learn More
Brady™ Keyed Different Safety Padlocks
Product Code. 15693487
Change view
Click to view available options
Color:
Black
Green
Orange
Red
Yellow
Unit Size:
Pack of 6
Product Code. Color unitSize
15693487 Red Pack of 6
11896043 Black Pack of 6
15643497 Black Pack of 6
11816053 Green Pack of 6
15613497 Green Pack of 6
15633497 Orange Pack of 6
11826053 Orange Pack of 6
11806053 Yellow Pack of 6
15623497 Yellow Pack of 6
11886043 Red Pack of 6
10 options
Product Code. 15693487
Brand: Brady™ 051339
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Brady™ Safety Padlocks are the ultimate locks for all safety and security needs, and are precision-machined for better tamper resistance.
  • Unique cylinder is insulated to protect workers from shocks when key is inserted
  • Six-pin precision-machined for more unique key cuts and better tamper resistance
  • Reserved, paracentric keyway provides optimal security
  • Special design prohibits the key from being released until the padlock shackle is closed
  • Locks are compact and lightweight and resist impact, temperature, chemicals and corrosion
  • Non-conductive, non-sparking body features a ribbed design for easier gripping and handling
  • One key supplied with each lock
  • Key retaining - key will not release until the padlock shackle is securely closed
Specifications
Product Type Safety Padlocks
Alternate Part Number 51339
Color Red
Material Reinforced Nylon
Description Safety Padlocks - Standard
Diameter (Metric) Shackle 6.5 mm
Dimensions (L x W) 38.50 x 86.50 mm
Height (Metric) 38 mm
Height (Metric) Shackle 38.00 mm
Material (Shackle) Hardened Steel
No. of Pins 6
Quantity 1 Ea.
Width (Metric) 33 mm