Learn More
Brady™ Keyed Different Safety Padlocks
Description
- Unique cylinder is insulated to protect workers from shocks when key is inserted
- Six-pin precision-machined for more unique key cuts and better tamper resistance
- Reserved, paracentric keyway provides optimal security
- Special design prohibits the key from being released until the padlock shackle is closed
- Locks are compact and lightweight and resist impact, temperature, chemicals and corrosion
- Non-conductive, non-sparking body features a ribbed design for easier gripping and handling
- One key supplied with each lock
- Key retaining - key will not release until the padlock shackle is securely closed
Specifications
Specifications
| Product Type | Safety Padlocks |
| Alternate Part Number | 51339 |
| Color | Red |
| Material | Reinforced Nylon |
| Description | Safety Padlocks - Standard |
| Diameter (Metric) Shackle | 6.5 mm |
| Dimensions (L x W) | 38.50 x 86.50 mm |
| Height (Metric) | 38 mm |
| Height (Metric) Shackle | 38.00 mm |
| Material (Shackle) | Hardened Steel |
| Show More |