Learn More
Thermo Scientific™ Accessories for Digital Vortex Mixers
Product Code. 16514812 Shop All Thermo Scientific Products
Click to view available options
Description:
12 x 15 mm dia. Tube Holder
13 x 12 mm dia. Tube Holder
19 x 10 mm dia.Tube Holder
4 x 26 mm dia.Tube Holder
7 x 20 mm dia. Tube Holder
Flat bottom vessel platform
Microplate Tray
Rubber Platform
Dimensions (L x W):
136.1 x 97.3 mm
82 x 16.5 mm
87.5 x 10 mm
87.5 x 40 mm
87.5 x 55 mm
Material:
Foam, plastic
Plastic
Rubber
Unit Size:
Each
Product Code. 16514812
Brand: Thermo Scientific™ 88882126
Only null left
Add to basket
Add to basket
Get assorted accessories for Digital Vortex Mixers
Assorted accessories for Digital Vortex Mixers
Service Plan
Standard Thermo warranty guideline, provide repair service and free spare part within warranty
Warranty
2 years
TRUSTED_SUSTAINABILITY
Specifications
Tube Size 20 mm dia.
Product Type Tube Holder
Material Foam, plastic
Dimensions (L x W) 87.5 x 55 mm
Color Black
Description 7 x 20 mm dia. Tube Holder
For Use With (Equipment) Digital Vortex mixer