Learn More
Fisherbrand™ Rubber Pipette Bulbs for Bulb or Cylindrical Pipettes
Excellent for all media that do not attack rubber. Fisherbrand™ Pipette Bulbs come with three glassball valves, adjustable by finger pressure.
Brand:  Fisherbrand™
Additional Details : Weight : 0.04320kg
Product Code. 11562672
  • 79.05 NOK / Each
Estimated Shipment: 27-05-2024
Description
  • Available in a range of colors
  • With ca. 60mL volume suitable for pipettes from 2 to 50mL
  • Pipette media must not enter pipette filler; it may cause valve malfunction