Learn More
Saint Gobain Life Sciences Rubber Gas Sample Bag
Product Code. 11754498
Change view
Click to view available options
Capacity (Metric):
0.5 L
2 L
5 L
10 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
11754498 5 L Each
11744498 2 L Each
11724498 0.5 L Each
11764498 10 L Each
4 options
Product Code. 11754498
Brand: Saint Gobain Life Sciences 5607076
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these bags for gas sample analysis. Saint Gobain Life Sciences Rubber Gas Sample Bags feature a cylindrical shape and an ebonite tube and tap.
  • Bags are individually packaged
  • Available in packs of ten
  • Foil thickness: 5/10mm
  • Custom sizes and configurations available on request
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 5 L
Includes Faucet
Material Rubber
Product Type Gas Sampling Bag