Learn More
Abeba™ Rubber 9252 Shoes
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather
- Black
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Leather insole
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20347:2012 OB A, FO SRC/li |
| Color | Black |
| Material | Leather |
| Size | 40 |
| Sole Features | Slip-resistant |
| Sole Material | Rubber |
| Style | 9252 |