Learn More
Abeba™ Rubber 9250 Shoes
Product Code. 15425723
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. Size unitSize
15425723 43 1 pair
15445713 36 1 pair
15455713 37 1 pair
15465713 38 1 pair
15475713 39 1 pair
15485713 40 1 pair
15495713 41 1 pair
15415723 42 1 pair
15435723 44 1 pair
15673208 45 1 pair
15683208 46 1 pair
15693208 47 1 pair
12 options
Product Code. 15425723
Brand: Abeba™ 9250/43
Only null left
Add to basket
Add to basket
Acid and base resistant. Rubber sole provides high slip resistance.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Dark blue
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC/li
Color Blue
Material Leather
Size 43
Sole Features Slip-resistant
Sole Material Rubber
Style 9250