Learn More
Abeba™ Rubber 5150 Shoes
Product Code. 15691828
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15691828 44 1 pair
15611828 36 1 pair
15621828 37 1 pair
15631828 38 1 pair
15641828 39 1 pair
15651828 40 1 pair
15661828 41 1 pair
15671828 42 1 pair
15681828 43 1 pair
15601838 45 1 pair
15611838 46 1 pair
15621838 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15691828 Supplier Abeba™ Supplier No. 5150/44
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 12-11-2026
ESD dissipative shoe is acid and base resistant.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Dark blue
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Leather top layer
  • Adjustable instep area
  • Lining microfiber, respiration absorbing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color Blue
Liner Material Microfiber
Material Leather
Size 44
Sole Features Slip-resistant
Sole Material Rubber
Style 5150