Learn More
Abeba™ Rubber 5100 Shoes
Product Code. 15691808
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15691808 36 1 pair
15601818 37 1 pair
15611818 38 1 pair
15621818 39 1 pair
15631818 40 1 pair
15641818 41 1 pair
15651818 42 1 pair
15661818 43 1 pair
15671818 44 1 pair
15681818 45 1 pair
15691818 46 1 pair
15601828 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 15691808 Supplier Abeba™ Supplier No. 5100/36
Only null left
Add to basket
Add to basket
ESD dissipative shoe is acid and base resistant.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather white
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Leather top layer
  • Adjustable instep area
  • Lining microfiber, respiration absorbing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color White
Liner Material Microfiber
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Rubber
Style 5100