Learn More
Abeba™ Rubber 5100 Shoes
Product Code. 15681818
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. Size unitSize
15681818 45 1 pair
15691808 36 1 pair
15601818 37 1 pair
15611818 38 1 pair
15621818 39 1 pair
15631818 40 1 pair
15641818 41 1 pair
15651818 42 1 pair
15661818 43 1 pair
15671818 44 1 pair
15691818 46 1 pair
15601828 47 1 pair
12 options
Product Code. 15681818
Brand: Abeba™ 5100/45
Only null left
Add to basket
Add to basket
ESD dissipative shoe is acid and base resistant.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather white
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Leather top layer
  • Adjustable instep area
  • Lining microfiber, respiration absorbing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color White
Liner Material Microfiber
Material Leather
Size 45
Sole Features Slip-resistant
Sole Material Rubber
Style 5100