missing translation for 'onlineSavingsMsg'
Learn More

RU 24969, MedChemExpress

Product Code. p-200101557
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493157

Brand: Medchem Express HY16688

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RU 24969 is a preferential 5-HT1B agonist, with a Ki of 0.38 nM, but also displays appreciable affinity for the 5-HT1A receptor (Ki=2.5 nM), and has low affinity for other receptor sites in the brain. RU 24969 could decrease fluid consumption and increase forward locomotion.

RU 24969 is a preferential 5-HT1B agonist, with a Ki of 0.38 nM, but also displays appreciable affinity for the 5-HT1A receptor (Ki=2.5 nM), and has low affinity for other receptor sites in the brain. RU 24969 could decrease fluid consumption and increase forward locomotion.

Specifications

Chemical Name or Material RU 24969
CAS 66611-26-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H16N2O
Quantity 50 mg
Solubility Information DMSO : ≥ 30 mg/mL (131.41 mM)
SMILES COC1=CC=C2C(C(C3=CCNCC3)=CN2)=C1
Molecular Weight (g/mol) 228.29
Formula Weight 228.29
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.