Learn More
RTI-51 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (1R,2S,3S,5S)-3-(4-Bromophenyl)-8-methyl-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester Hydrochloride |
| CAS | 1391052-88-2 |
| Molecular Formula | C16H21BrClNO2 |
| Quantity | 2.5 mg |
| Synonym | [1R-(exo,exo)]-3-(4-Bromophenyl)-8-methyl-8-azabicyclo[3.2.1]octane-2-carboxylic Acid Methyl Ester Hydrochloride, RTI 4229-51 Hydrochloride, RTI-51 Hydrochloride |
| InChI Formula | InChI=1S/C16H20BrNO2.ClH/c1-18-12-7-8-14(18)15(16(19)20-2)13(9-12)10-3-5-11(17)6-4-10;/h3-6,12-15H,7-9H2,1-2H3;1H/t12-,13+,14+,15-;/m0./s1 |
| SMILES | Cl.COC(=O)[C@@H]1[C@H]2CC[C@@H](C[C@@H]1c3ccc(Br)cc3)N2C |
| IUPAC Name | methyl (1S,3S,4S,5R)-3-(4-bromophenyl)-8-methyl-8-azabicyclo[3.2.1]octane-4-carboxylate;hydrochloride |
| Molecular Weight (g/mol) | 374.7 |
| Formula Weight | 373.0444 |