missing translation for 'onlineSavingsMsg'
Learn More

RSPH9 Antibody - BSA Free, Novus Biologicals™

Product Code. 18667721 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
Product Code. Quantity unitSize
18667721 0.02 mL 0.02mL
18663591 0.1 mL 0.01mL
2 options
This item is not returnable. View return policy

Product Code. 18667721

Brand: Novus Biologicals NBP2944790.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RSPH9 Polyclonal antibody specifically detects RSPH9 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RSPH9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias C6orf206, chromosome 6 open reading frame 206, CILD12, FLJ30845, mitochondrial ribosomal protein S18A-like 1, MRPS18AL1, radial spoke head 9 homolog (Chlamydomonas), radial spoke head protein 9 homolog
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-276 of human RSPH9 (NP_689945.2). MDADSLLLSLELASGSGQGLSPDRRASLLTSLMLVKRDYRYDRVLFWGRILGLVADYYIAQGLSEDQLAPRKTLYSLNCTEWSLLPPATEEMVAQSSVVKGRFMGDPSYEYEHTELQKVNEGEKVFEEEIVVQIKEETRLVSVIDQIDKAVAIIPRGALFKTPFGPTHVNRTFEGLSLSEAKKLSSYFHFREPVELKNKTLLEKADLDPSLDFMDSLEHDIPKGSWSIQMERGNALVVLRSLLWPGLTFYHAPRTKNYGYVYVGTGEKNMDLPFML
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 221421
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.