missing translation for 'onlineSavingsMsg'
Learn More

RSL24D1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25190-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893745

  • 3468.15 NOK / 150µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RSL24D1
Polyclonal
Unconjugated
RSL24D1
C15orf15, HRP L30 iso, L30, Ribosomal protein L24 like, RLP24, RPL24, RPL24L, RSL24D1, TVAS3
Rabbit
Antigen Affinity Chromatography
RUO
51187
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UHA3
RSL24D1
RSL24D1 Fusion Protein Ag18195
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers