Learn More
[(RS)-(Diphenylmethyl)sulfinyl]acetic Acid, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
[(RS)-(Diphenylmethyl)sulfinyl]acetic Acid
Specifications
Specifications
| Chemical Name or Material | Modafinil acid |
| CAS | 63547-24-0 |
| Analyte or Component Names | Modafinil acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H14 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Modafinil Imp. A (EP), Modafinil USP Related Compound A, Modafinil USP RC A, [(RS)-(Diphenylmethyl)sulfinyl]acetic Acid, Modafinil Carboxylate, Modafinil acid, Modafinil Related Compound A, Modafinil Impurity A |
| InChI Formula | InChI=1S/C15H14O3S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H,16,17) |
| SMILES | OC(=O)CS(=O)C(c1ccccc1)c2ccccc2 |
| IUPAC Name | 2-benzhydrylsulfinylacetic acid |
| Show More |