Learn More
RS-246204, MedChemExpress
Product Code. 16464638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464638 10 mg 10mg
16455476 100 mg 100mg
16443176 25 mg 25mg
16437357 5 mg 5mg
16400937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16464638 Supplier Medchem Express Supplier No. HY112484
MedChemExpress RS-246204 is a R-spondin-1 substitute compound that is able initiate small intestinal organoids without the use of the R-spondin-1 protein.
RS-246204 is a R-spondin-1 substitute compound that is able initiate small intestinal organoids without the use of the R-spondin-1 protein.
Specifications
Chemical Name or Material RS-246204
CAS 878451-87-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H13N5O2S
Quantity 10 mg
Solubility Information DMSO : 5 mg/mL (14.73 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N1)N(C)C2=C(N(C)C(SC3=NC4=CC=CC=C4C=C3)=N2)C1=O
Molecular Weight (g/mol) 339.37
Formula Weight 339.37
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death