missing translation for 'onlineSavingsMsg'
Learn More

RS-246204, MedChemExpress

Product Code. 16400937
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16400937 50 mg 50mg
16464638 10 mg 10mg
16455476 100 mg 100mg
16443176 25 mg 25mg
16437357 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16400937 Supplier Medchem Express Supplier No. HY112484

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RS-246204 is a R-spondin-1 substitute compound that is able initiate small intestinal organoids without the use of the R-spondin-1 protein.

RS-246204 is a R-spondin-1 substitute compound that is able initiate small intestinal organoids without the use of the R-spondin-1 protein.

Specifications

Chemical Name or Material RS-246204
CAS 878451-87-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H13N5O2S
Quantity 50 mg
Solubility Information DMSO : 5 mg/mL (14.73 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N1)N(C)C2=C(N(C)C(SC3=NC4=CC=CC=C4C=C3)=N2)C1=O
Molecular Weight (g/mol) 339.37
Formula Weight 339.37
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.