Learn More
RRN3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16844465 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16844465 20 μL 20µL
16834465 150 μL 150µL
2 options
Product Code. 16844465
Brand: Proteintech 259181AP20UL
Rabbit Polyclonal Antibody
The protein encoded by this gene forms a complex with two other proteins, NPL4 and VCP, that is necessary for the degradation of ubiquitinated proteins. In addition, this complex controls the disassembly of the mitotic spindle and the formation of a closed nuclear envelope after mitosis. Mutations in this gene have been associated with Catch 22 syndrome as well as cardiac and craniofacial defects. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
Antigen RRN3
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RRN3
Gene Accession No. B2RS91, Q9NYV6
Gene Alias RRN3, TIF IA, TIFIA
Gene Symbols RRN3
Host Species Rabbit
Immunogen RRN3 Fusion Protein Ag23207
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106298, 54700
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG