Learn More
RRM2 Mouse anti-Human, Unconjugated, Clone: 2A9A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67006-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889365
  • 1611.84 NOK / 20µL
Estimated Shipment: 23-05-2024
Description
The RRM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RRM2. This antibody recognizes human antigen. The RRM2 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Specifications
Specifications
RRM2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunoprecipitation, Immunofluorescence, ELISA
2A9A7
Unconjugated
20 μL
6241
IgG1