missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RREB1-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
The RREB1-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human RREB1-Specific. This antibody recognizes human antigen. The RREB1-Specific antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
| Antigen | RREB1-Specific |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | 187 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 6239 |
| Target Species | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?