Learn More
Invitrogen™ RRAGB Polyclonal Antibody
GREENER_CHOICE
Product Code. 15596952
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15596952 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15596952 Supplier Invitrogen™ Supplier No. PA545812
Rabbit Polyclonal Antibody
Peptide sequence: IDIFTSNTYV MVVMSDPSIP SAATLINIRN ARKHFEKLER VDGPKQCLLM Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 93%.
Ras-homologous GTPases constitute a large family of signal transducers that alternate between an activated, GTP-binding state and an inactivated, GDP-binding state. These proteins represent cellular switches that are operated by GTP-exchange factors and factors that stimulate their intrinsic GTPase activity. All GTPases of the Ras superfamily have in common the presence of six conserved motifs involved in GTP/GDP binding, three of which are phosphate-/magnesium-binding sites (PM1-PM3) and three of which are guanine nucleotide-binding sites (G1-G3). Transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Antigen RRAGB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RRAGB
Gene Accession No. Q5VZM2
Gene Alias bA465E19.1; GTP-binding protein ragB; LOC100724885; Rag B; RAGB; Ras related GTP binding B; Ras-related GTP binding B; ras-related GTP-binding protein A; ras-related GTP-binding protein B; ras-related GTP-binding protein B; RGD:619805}; RRAGB; rragB {ECO:0000312
Gene Symbols RRAGB
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RRAGB (aa 324-373).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10325
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG