missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS4Y1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17296-1-AP-150UL
This item is not returnable.
View return policy
Description
As a homolog with RPS4X, the antibody can recognize endogenous level of RPS4Y and RPS4X, at 29-32 kDa.
Cytoplasmic ribosomes, organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes ribosomal protein S4, a component of the 40S subunit. Ribosomal protein S4 is the only ribosomal protein known to be encoded by more than one gene, namely this gene and ribosomal protein S4, X-linked (RPS4X). The 2 isoforms encoded by these genes are not identical, but are functionally equivalent. Ribosomal protein S4 belongs to the S4E family of ribosomal proteins. It has been suggested that haploinsufficiency of the ribosomal protein S4 genes plays a role in Turner syndrome; however, this hypothesis is controversial. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Specifications
| RPS4Y1 | |
| Polyclonal | |
| Unconjugated | |
| RPS4X | |
| RPS4Y, RPS4Y1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100362640, 20102, 6191, 6192 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22090, P62701, P62702, P62703 | |
| RPS4X, RPS4Y1 | |
| RPS4Y1 Fusion Protein Ag11176 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction