missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16946-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.Specifications
RPS21 | |
Polyclonal | |
Unconjugated | |
RPS21 | |
40S ribosomal protein S21, ribosomal protein S21, RPS21 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6227, 66481, 81775 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05765, P63220, Q9CQR2 | |
RPS21 | |
RPS21 Fusion Protein Ag10557 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |