missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RPS17 Recombinant Protein

Brand:  Abnova™ H00006218-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190372

  • 4115.00 NOK / 10µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH09407
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
40.59
8
Glutathione Sepharose 4 Fast Flow
10 ug
MGRVRTKTVKKAARVIIEKYYTRLGNDFHTNKRVCEEIAIIPSKKLRNKIAGYVTHLMKRIQRGPVRGISIKLQEEERERRDNYVPEVSALDQEIIEVDPDTKEMLKLLDFGSLSNLQVTQPTVGMNFKTPRGPV
DBA4/MGC72007/RPS17L1/RPS17L2
RPS17
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
6218
RPS17 (Human) Recombinant Protein (P02)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPS17
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers